N-(9-((2R,3R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-3-hydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide 95%

Supplier: Ambeed
Warning

Synonyms: 5'-DMT-3'-TBDMS-Bz-rA

A173995-1G A173995-250MG A173995-5G A173995-100MG
77163-670EA 50 USD
77163-670 77163-672 77567-564 77892-151
N-(9-((2R,3R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-3-hydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide 95%
N-(9-((2R,3R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-3-hydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide

Precaución: For research use only.

Formula: C₄₄H₄₉N₅O₇Si
MW: 787.97 g/mol
MDL Number: MFCD00274103
CAS Number: 81256-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR