(S)-(-)-3-Aminoquinuclidine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Quinuclidin-3-amine dihydrochloride, (S)-Quinuclidin-3-amine 2HCl, (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

A173742-10G A173742-250MG A173742-5G A173742-100G A173742-1G A173742-25G
77191-488EA 49 USD
77191-488 77191-490 77191-492 77615-454 77746-145 77746-146
(S)-(-)-3-Aminoquinuclidine dihydrochloride 97%
(S)-(-)-3-Aminoquinuclidine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂
MW: 199.1213 g/mol
MDL Number: MFCD00191753
CAS Number: 119904-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR