1-Benzyloxycarbonyl-3-piperidinemethanol 96%

Supplier: Ambeed
Warning

Synonyms: Benzyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(Hydroxymethyl)piperidine, N-CBZ protected

A173656-10G A173656-1G A173656-25G A173656-5G
77163-662EA 55 USD
77163-662 77163-664 77163-666 77163-668
1-Benzyloxycarbonyl-3-piperidinemethanol 96%
1-Benzyloxycarbonyl-3-piperidinemethanol

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MDL Number: MFCD03407298
CAS Number: 39945-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR