AZD-3965 98%

Supplier: Ambeed
Warning

Synonyms: AZD3965, (S)-5-(4-Hydroxy-4-methylisoxazolidine-2-carbonyl)-1-isopropyl-3-methyl-6-((5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione, (S)-5-(4-hydroxy-4-methylisoxazolidine-2-carbonyl)-1-isopropyl-3-methyl-6-((3-methyl-5-(trifluoromethyl)-1H-pyrazol-4-yl)methyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione

A173643-10MG A173643-1MG A173643-2MG A173643-5MG
77163-654EA 112.27 USD
77163-654 77163-656 77163-658 77163-660
AZD-3965 98%
AZD-3965

Precaución: for Research Use Only

Formula: C₂₁H₂₄F₃N₅O₅S
MW: 515.51 g/mol
MDL Number: MFCD28963899
CAS Number: 1448671-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR