3-(2-Thienyl)-D-alanine 95%

Supplier: Ambeed
Warning

Synonyms: H-β-(2-Thienyl)-D-Ala-OH, (R)-2-Amino-3-(thiophen-2-yl)propanoic acid, H-D-3-(2-Thienyl)-Ala-OH

A173569-250MG A173569-1G A173569-5G
77191-486EA 14.42 USD
77191-486 77746-143 77746-144
3-(2-Thienyl)-D-alanine 95%
3-(2-Thienyl)-D-alanine

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.214 g/mol
MDL Number: MFCD00077166
CAS Number: 62561-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR