N-(6-Bromohexyl)phthalimide 96%

Supplier: Ambeed
Warning

A172673-100G A172673-1G A172673-25G A172673-5G
77191-472EA 356.38 USD
77191-472 77191-474 77191-476 77191-478
N-(6-Bromohexyl)phthalimide 96%
N-(6-Bromohexyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₄H₁₆BrNO₂
MW: 310.19 g/mol
MDL Number: MFCD00023098
CAS Number: 24566-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR