Benzofuran-6-ol 95%

Supplier: Ambeed
Warning

A172430-100MG A172430-1G
77191-454EA 68 USD
77191-454 77191-456
Benzofuran-6-ol 95%
Benzofuran-6-ol

Precaución: for Research Use Only

Formula: C₈H₆O₂
MDL Number: MFCD08669481
CAS Number: 13196-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR