CAY10683 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl (4-oxo-4-(phenethylamino)butyl)carbamate, EthylN-[4-oxo-4-(2-phenylethylamino)butyl]carbamate, Santacruzamate A
A172133-10MG
A172133-1MG
A172133-5MG
77163-596EA
47.99
USD
77163-596
77163-598
77163-600
CAY10683 98%
CAY10683
Precaución: for Research Use Only
Formula:
C₁₅H₂₂N₂O₃ |
CAS Number:
1477949-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...