1,1,4,4,7-Pentamethylisochroman 97%

Supplier: Ambeed
Warning

A171963-100MG A171963-250MG
77191-440EA 27 USD
77191-440 77191-442
1,1,4,4,7-Pentamethylisochroman 97%
1,1,4,4,7-Pentamethylisochroman

Precaución: for Research Use Only

Formula: C₁₄H₂₀O
MDL Number: MFCD18807174
CAS Number: 786690-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR