FPS-ZM1 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-N-cyclohexyl-N-(phenylmethyl)benzamide, N-Benzyl-4-chloro-N-cyclohexylbenzamide

A171795-10MG A171795-1G A171795-1MG A171795-50MG A171795-5MG A171795-100MG
77191-430EA 13.39 USD
77191-430 77191-432 77191-434 77191-436 77191-438 77746-137
FPS-ZM1 98%
FPS-ZM1

Precaución: for Research Use Only

Formula: C₂₀H₂₂ClNO
MW: 327.85 g/mol
MDL Number: MFCD22378757
CAS Number: 945714-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR