1-(2-Methoxy-5-methylphenyl)ethanone 95%

Supplier: Ambeed
Warning

A171197-100MG A171197-1G A171197-250MG A171197-5G
77191-414EA 13.39 USD
77191-414 77191-416 77191-418 77191-420
1-(2-Methoxy-5-methylphenyl)ethanone 95%
1-(2-Methoxy-5-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
MDL Number: MFCD00156674
CAS Number: 20628-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR