Methyl-5-methyl-2-furoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-methylfuran-2-carboxylate
A171077-10G
A171077-1G
A171077-250MG
A171077-5G
A171077-25G
77191-402EA
58
USD
77191-402
77191-404
77191-406
77191-408
77746-135
Methyl-5-methyl-2-furoate 97%
Methyl-5-methyl-2-furoate
Precaución: for Research Use Only
Formula:
C₇H₈O₃ |
MDL Number:
MFCD04116374 CAS Number: 2527-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...