4-Fluoro-2-methylbenzo[d]oxazole 97%

Supplier: Ambeed
Warning

A170776-100MG A170776-10G A170776-1G A170776-250MG
77191-390EA 21.63 USD
77191-390 77191-392 77191-394 77191-396
4-Fluoro-2-methylbenzo[d]oxazole 97%
4-Fluoro-2-methylbenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD25978067
CAS Number: 1804412-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR