1-Methyl-1H-pyrazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyrazole-3-carboxylic acid

A170768-100MG A170768-1G A170768-250MG A170768-5G A170768-10G A170768-100G
77163-566EA 5.15 USD
77163-566 77163-568 77163-570 77567-510 77611-668 77683-391
1-Methyl-1H-pyrazole-3-carboxylic acid 98%
1-Methyl-1H-pyrazole-3-carboxylic acid

Precaución: For research use only.

Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
MDL Number: MFCD00464254
CAS Number: 25016-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR