4-Fluoro-1-isopropyl-2-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A170573-100MG A170573-1G A170573-250MG A170573-25G A170573-5G
77163-548EA 20.6 USD
77163-548 77163-550 77163-552 77163-554 77683-390
4-Fluoro-1-isopropyl-2-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole 95%
4-Fluoro-1-isopropyl-2-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole

Precaución: For research use only.

Formula: C₁₇H₂₄BFN₂O₂
MW: 318.19 g/mol
MDL Number: MFCD25977327
CAS Number: 1231930-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR