4-Chloro-2-fluoro-1,1'-biphenyl 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-chloro biphenyl

A170401-1G A170401-250MG
77191-382EA 156.56 USD
77191-382 77191-384
4-Chloro-2-fluoro-1,1'-biphenyl 95%
4-Chloro-2-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈ClF
MW: 206.64 g/mol
MDL Number: MFCD20267653
CAS Number: 39224-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR