6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

A170209-100MG A170209-1G A170209-250MG
77163-528EA 44 USD
77163-528 77163-530 77163-532
6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine 97%
6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₃BrF₃N₃
MW: 266.018 g/mol
MDL Number: MFCD11111700
CAS Number: 1166819-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR