N-(4-Fluoro-2-methoxy-5-nitrophenyl)-4-(1-methyl-1H-indol-3-yl)pyrimidin-2-amine 95%
Supplier: Ambeed
|
Warning
|
A170176-10G
A170176-1G
A170176-250MG
A170176-25G
A170176-5G
77163-518EA
100.94
USD
77163-518
77163-520
77163-522
77163-524
77163-526
N-(4-Fluoro-2-methoxy-5-nitrophenyl)-4-(1-methyl-1H-indol-3-yl)pyrimidin-2-amine 95%
N-(4-Fluoro-2-methoxy-5-nitrophenyl)-4-(1-methyl-1H-indol-3-yl)pyrimidin-2-amine
Precaución: for Research Use Only
Formula:
C₂₀H₁₆FN₅O₃ MW: 393.37 g/mol |
MDL Number:
MFCD28167946 CAS Number: 1421372-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...