11-Mercaptoundecan-1-ol 97%

Supplier: Ambeed
Danger

A169910-1G A169910-250MG A169910-5G
77191-374EA 78 USD
77191-374 77191-376 77593-050
11-Mercaptoundecan-1-ol 97%
11-Mercaptoundecan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₂₄OS
MDL Number: MFCD00239433
CAS Number: 73768-94-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR