3-Chlorophenyl acetylene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-3-ethynylbenzene, 3-Chlorophenylacetylene, (3-Chlorophenyl)acetylene

A169165-100G A169165-10G A169165-1G A169165-25G A169165-5G
77191-356EA 199 USD
77191-356 77191-358 77191-360 77191-362 77191-364
3-Chlorophenyl acetylene 98%
3-Chlorophenyl acetylene

Precaución: for Research Use Only

Formula: C₈H₅Cl
MW: 136.578 g/mol
MDL Number: MFCD03839984
CAS Number: 766-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR