Mono-(6-amino-6-deoxy)-β-cyclodextrin 95%

Supplier: Ambeed
Warning

A168954-1G A168954-250MG
77163-442EA 255.44 USD
77163-442 77163-444
Mono-(6-amino-6-deoxy)-β-cyclodextrin 95%
Mono-(6-amino-6-deoxy)-β-cyclodextrin

Precaución: for Research Use Only

Formula: C₄₂H₇₁NO₃₄
MW: 1133.99 g/mol
MDL Number: MFCD03452964
CAS Number: 29390-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR