4-(4-Chlorophenyl)pyridine 95%

Supplier: Ambeed
Warning

A168312-1G A168312-250MG
77163-410EA 186.43 USD
77163-410 77163-412
4-(4-Chlorophenyl)pyridine 95%
4-(4-Chlorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈ClN
MW: 189.64 g/mol
MDL Number: MFCD00956749
CAS Number: 5957-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR