2-Amino-6-bromo-3-methylpyridine 97%

Supplier: Ambeed
Warning

A168268-1G A168268-250MG A168268-25G A168268-5G
77163-402EA 15 USD
77163-402 77163-404 77163-406 77163-408
2-Amino-6-bromo-3-methylpyridine 97%
2-Amino-6-bromo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MDL Number: MFCD17014961
CAS Number: 89466-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR