4-(tert-Butyl)-2-chloropyridine 97%

Supplier: Ambeed
Danger

A167518-10G A167518-5G A167518-1G
77163-380EA 99.91 USD
77163-380 77567-444 77611-634
4-(tert-Butyl)-2-chloropyridine 97%
4-(tert-Butyl)-2-chloropyridine

Precaución: For research use only.

Formula: C₉H₁₂ClN
MW: 169.65 g/mol
MDL Number: MFCD12131129
CAS Number: 81167-60-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR