3-(Adamantan-1-yl)-6-bromonaphthalen-2-ol 95%

Supplier: Ambeed
Warning

A167276-100MG A167276-1G A167276-250MG
77191-314EA 43 USD
77191-314 77191-316 77191-318
3-(Adamantan-1-yl)-6-bromonaphthalen-2-ol 95%
3-(Adamantan-1-yl)-6-bromonaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₂₀H₂₁BrO
MDL Number: MFCD13182367
CAS Number: 128272-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR