2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Formyl-4-(trifluoromethyl)phenylboronic acid pinacol ester
A167187-100MG
A167187-1G
A167187-250MG
77191-308EA
35.02
USD
77191-308
77191-310
77191-312
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzaldehyde 98%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₆BF₃O₃ MW: 300.08 g/mol |
MDL Number:
MFCD18383807 CAS Number: 1219936-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...