6-Amino-2-chloronicotinonitrile 95%

Supplier: Ambeed
Warning

A167074-100MG A167074-1G A167074-250MG
77163-378EA 183.34 USD
77163-378 77683-368 77683-369
6-Amino-2-chloronicotinonitrile 95%
6-Amino-2-chloronicotinonitrile

Precaución: For research use only.

Formula: C₆H₄ClN₃
MW: 153.57 g/mol
MDL Number: MFCD18802573
CAS Number: 53554-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR