1-Amino-1-phenylacetone hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-1-phenylpropan-2-one hydrochloride

A166862-1G A166862-250MG
77191-300EA 344.02 USD
77191-300 77191-302
1-Amino-1-phenylacetone hydrochloride 95%
1-Amino-1-phenylacetone hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MW: 185.66 g/mol
MDL Number: MFCD01934460
CAS Number: 3904-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR