5-(Methoxycarbonyl)pyridine-3-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-borononicotinate, Methyl 5-boronopyridine-3-carboxylate, (5-(Methoxycarbonyl)pyridin-3-yl)boronic acid

A166468-100MG A166468-1G A166468-250MG A166468-5G
77191-284EA 29 USD
77191-284 77191-286 77191-288 77191-290
5-(Methoxycarbonyl)pyridine-3-boronic acid 97%
5-(Methoxycarbonyl)pyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₄
MW: 180.954 g/mol
MDL Number: MFCD07363742
CAS Number: 871329-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR