1-(4-Nitrophenyl)-2-pyrrolidinone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Nitrophenyl)pyrrolidin-2-one

A166416-100MG A166416-1G A166416-250MG
77191-278EA 21.63 USD
77191-278 77191-280 77191-282
1-(4-Nitrophenyl)-2-pyrrolidinone 98%
1-(4-Nitrophenyl)-2-pyrrolidinone

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MW: 206.2 g/mol
MDL Number: MFCD00138492
CAS Number: 13691-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR