3-Fluoro-5-(trifluoromethyl)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-fluoro-5-(trifluoromethyl)benzaldehyde

A166407-10G A166407-1G A166407-5G
77191-272EA 49.44 USD
77191-272 77191-274 77191-276
3-Fluoro-5-(trifluoromethyl)benzaldehyde 97%
3-Fluoro-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MW: 192.11 g/mol
MDL Number: MFCD00061312
CAS Number: 188815-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR