1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3S,4R,5R,6R)-6-(Acetoxymethyl)-3-hydroxytetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose

A166077-1G A166077-250MG A166077-5G A166077-25G
77191-262EA 35 USD
77191-262 77191-264 77615-380 77746-121
1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose 97%
1,3,4,6-Tetra-O-acetyl-beta-D-mannopyranose

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₁₀
MDL Number: MFCD00012354
CAS Number: 18968-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR