Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate 97%

Supplier: Ambeed
Warning

Synonyms: Sodium 5-Isopropyl-3,8-dimethylazulene-1-sulphonate, Sodium Guaiazulenesulphonate, 7-Isopropyl-1,4-dimethyl-azulene-3-sulphonic acid sodium salt, 5-Isopropyl-3,8-dimethylazulene-1-sulphonic acid sodium salt, Guaiazulenesulphonic acid sodium salt, Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulphonate

A165846-1G A165846-250MG A165846-25G A165846-5G
77191-254EA 17.51 USD
77191-254 77191-256 77191-258 77191-260
Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate 97%
Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NaO₃S
MW: 300.35 g/mol
MDL Number: MFCD00866167
CAS Number: 6223-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR