1-(4-Methylpyrimidin-5-yl)ethanone 95%

Supplier: Ambeed
Warning

A165406-100MG A165406-250MG
77191-246EA 84 USD
77191-246 77191-248
1-(4-Methylpyrimidin-5-yl)ethanone 95%
1-(4-Methylpyrimidin-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD09263922
CAS Number: 87379-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR