1-Boc-indole-3-boronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1-(tert-Butoxycarbonyl)-1H-indol-3-yl)boronic acid
A165202-10G
A165202-1G
A165202-250MG
A165202-25G
A165202-5G
77191-234EA
98
USD
77191-234
77191-236
77191-238
77191-240
77191-242
1-Boc-indole-3-boronic acid 97%
1-Boc-indole-3-boronic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BNO₄ MW: 261.0814 g/mol |
MDL Number:
MFCD06656257 CAS Number: 181365-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...