2-Bromo-1-ethylpyridinium tetrafluoroborate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate
A164811-10G
A164811-1G
A164811-250MG
A164811-25G
A164811-5G
77191-222EA
63.86
USD
77191-222
77191-224
77191-226
77191-228
77191-230
2-Bromo-1-ethylpyridinium tetrafluoroborate 98%
2-Bromo-1-ethylpyridinium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₇H₉BBrF₄N MW: 273.86 g/mol |
MDL Number:
MFCD00060046 CAS Number: 878-23-9 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...