Methyl 1,5-dimethyl-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

A164573-100MG A164573-1G A164573-250MG
77191-206EA 57 USD
77191-206 77191-208 77191-210
Methyl 1,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Methyl 1,5-dimethyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.17 g/mol
MDL Number: MFCD00044616
CAS Number: 73476-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR