3,4-Dichloro-2-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A164336-1G A164336-250MG
77191-200EA 59 USD
77191-200 77191-202
3,4-Dichloro-2-methoxybenzoic acid 95%
3,4-Dichloro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
MDL Number: MFCD28123955
CAS Number: 155382-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR