7-Bromo-6-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A163851-100MG A163851-1G A163851-250MG A163851-5G
77163-300EA 29 USD
77163-300 77163-302 77163-304 77163-306
7-Bromo-6-methyl-1H-indazole 97%
7-Bromo-6-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MDL Number: MFCD17168273
CAS Number: 1257535-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR