3-Bromo-4-iodobenzonitrile 97%

Supplier: Ambeed
Warning

A163717-100G A163717-1G A163717-25G A163717-5G A163717-10G
77163-290EA 271 USD
77163-290 77163-292 77163-294 77163-296 77163-298 77892-107
3-Bromo-4-iodobenzonitrile 97%
3-Bromo-4-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrIN
MDL Number: MFCD09800817
CAS Number: 1000577-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR