2-(Boc-amino)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl N-(2-pyridyl)carbamate, tert-Butyl pyridin-2-ylcarbamate

A163409-10G A163409-1G A163409-25G A163409-5G
77163-262EA 57.68 USD
77163-262 77163-264 77163-266 77163-268
2-(Boc-amino)pyridine 97%
2-(Boc-amino)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
MDL Number: MFCD03411622
CAS Number: 38427-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR