2,3,5,6,7,8-Hexahydrophthalazine-1,4-dione 97%

Supplier: Ambeed
Warning

A163044-100MG A163044-1G A163044-250MG A163044-5G
77163-248EA 5 USD
77163-248 77163-250 77163-252 77163-254
2,3,5,6,7,8-Hexahydrophthalazine-1,4-dione 97%
2,3,5,6,7,8-Hexahydrophthalazine-1,4-dione

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD27927305
CAS Number: 67279-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR