(R)-9-(2-Hydroxypropyl)adenine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-6-Amino-9-(2-hydroxypropyl)purine, (R)-1-(6-Amino-9H-purin-9-yl)propan-2-ol
A162737-100G
A162737-10G
A162737-1G
A162737-25G
A162737-500G
A162737-5G
77191-132EA
37
USD
77191-132
77191-134
77191-136
77191-138
77191-140
77191-142
(R)-9-(2-Hydroxypropyl)adenine 97%
(R)-9-(2-Hydroxypropyl)adenine
Precaución: for Research Use Only
Formula:
C₈H₁₁N₅O |
MDL Number:
MFCD07369451 CAS Number: 14047-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...