(6-Methoxy-pyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: (6-Methoxypyridin-2-yl)methanol

A162247-100MG A162247-1G A162247-250MG
77163-242EA 15.45 USD
77163-242 77163-244 77683-342
(6-Methoxy-pyridin-2-yl)methanol 97%
(6-Methoxy-pyridin-2-yl)methanol

Precaución: For research use only.

Formula: C₇H₉NO₂
MW: 139.16 g/mol
MDL Number: MFCD08235128
CAS Number: 63071-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR