Efinaconazole 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R)-2-(2,4-Difluorophenyl)-3-(4-methylenepiperidin-1-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, KP-13

A162236-100MG A162236-10MG A162236-1G A162236-1MG A162236-25MG A162236-50MG A162236-5G A162236-5MG A162236-250MG
77163-226EA 35.02 USD
77163-226 77163-228 77163-230 77163-232 77163-234 77163-236 77163-238 77163-240 77683-341
Efinaconazole 98%
Efinaconazole

Precaución: For research use only.

Formula: C₁₈H₂₂F₂N₄O
MW: 348.39 g/mol
MDL Number: MFCD00936407
CAS Number: 164650-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR