Spautin-1 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Fluoro-N-(4-fluorobenzyl)quinazolin-4-amine, 6-Fluoro-N-[(4-fluorophenyl)methyl]-4-quinazolinamine
A161476-10MG
A161476-1MG
A161476-25MG
A161476-5MG
A161476-50MG
77163-196EA
47.38
USD
77163-196
77163-198
77163-200
77163-202
77683-337
Spautin-1 98%
Spautin-1
Precaución: For research use only.
Formula:
C₁₅H₁₁F₂N₃ MW: 271.27 g/mol |
MDL Number:
MFCD16962292 CAS Number: 1262888-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...