3-Nitro-5-(trifluoromethyl)benzaldehyde 98%

Supplier: Ambeed
Warning

A160808-100MG A160808-1G A160808-250MG A160808-5G
77163-172EA 28 USD
77163-172 77163-174 77163-176 77163-178
3-Nitro-5-(trifluoromethyl)benzaldehyde 98%
3-Nitro-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₃
MW: 219.117 g/mol
MDL Number: MFCD11847795
CAS Number: 284047-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR