Methyl 2-bromoquinoline-4-carboxylate 95%

Supplier: Ambeed
Warning

A160601-100MG A160601-250MG A160601-5G A160601-1G
77191-064EA 50.47 USD
77191-064 77191-066 77191-068 77746-096
Methyl 2-bromoquinoline-4-carboxylate 95%
Methyl 2-bromoquinoline-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO₂
MW: 266.09 g/mol
MDL Number: MFCD26401251
CAS Number: 103502-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR