S-(Carboxymethyl)-L-cysteine 98%

Supplier: Ambeed
Warning

Synonyms: 3-[(Carboxymethyl)thio]alanine, Carbocisteine, Carbocysteine

A160278-100G A160278-25G A160278-500G A160278-5G A160278-1000G
77163-164EA 24.72 USD
77163-164 77163-166 77163-168 77163-170 77683-334
S-(Carboxymethyl)-L-cysteine 98%
S-(Carboxymethyl)-L-cysteine

Precaución: For research use only.

Formula: C₅H₉NO₄S
MW: 179.19 g/mol
MDL Number: MFCD00002614
CAS Number: 638-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR