3-Chloro-6-(3-nitrophenyl)pyridazine 95%

Supplier: Ambeed
Warning

A160020-100MG A160020-1G A160020-250MG
77191-044EA 72.1 USD
77191-044 77191-046 77191-048
3-Chloro-6-(3-nitrophenyl)pyridazine 95%
3-Chloro-6-(3-nitrophenyl)pyridazine

Precaución: for Research Use Only

Formula: C₁₀H₆ClN₃O₂
MW: 235.63 g/mol
MDL Number: MFCD09881851
CAS Number: 58059-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR